Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV34686

Sigma-Aldrich

Anti-PAX5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Paired box gene 5 (B-cell lineage specific activator)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

mouse, rabbit, guinea pig, human, dog, bovine, rat, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PAX5(7849)

General description

PAX5 is a transcription factor that regulates the differentiation and functions of B cells. It has been reported that Blimp-1-mediated PAX5 repression facilitates plasmacytic differentiation of B cells. Furthermore, PAX5 has been implicated in human B cell malignancies.
Rabbit Anti-PAX5 antibody recognizes human, mouse, rat, canine, zebrafish, and chicken PAX5.

Immunogen

Synthetic peptide directed towards the N terminal region of human PAX5

Application

Rabbit Anti-PAX5 antibody is suitable for western blot applications at a concentration of 2 μg/ml and for immunohistochemistry applications at 4-8 μg/ml.

Biochem/physiol Actions

The PAX5 gene is a member of the paired box (PAX) family of transcription factors. The central feature of this gene family is a novel, highly conserved DNA-binding motif, known as the paired box. The PAX proteins are important regulators in early development, and alterations in the expression of their genes are thought to contribute to neoplastic transformation. The PAX5 gene encodes the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 gene product may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis.

Sequence

Synthetic peptide located within the following region: CVSKILGRYYETGSIKPGVIGGSKPKVATPKVVEKIAEYKRQNPTMFAWE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuo-I Lin et al.
Molecular and cellular biology, 22(13), 4771-4780 (2002-06-08)
B-cell lineage-specific activator protein (BSAP), encoded by the Pax-5 gene, is critical for B-cell lineage commitment and B-cell development but is not expressed in terminally differentiated B cells. We demonstrate a direct connection between BSAP and B-lymphocyte-induced maturation protein 1
César Cobaleda et al.
Nature immunology, 8(5), 463-470 (2007-04-19)
The transcription factor Pax5 is essential for commitment of lymphoid progenitors to the B lymphocyte lineage. Pax5 fulfils a dual role by repressing B lineage 'inappropriate' genes and simultaneously activating B lineage-specific genes. This transcriptional reprogramming restricts the broad signaling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service