Las1L is a nucleolar protein required for cell proliferation and ribosome biogenesis. I has a molecular weight of 83 kDa.
Immunogen
The immunogen for anti-LAS1L antibody: synthetic peptide derected towards the middle region of human LAS1L
Application
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below. Immunoprecipitation (1 paper)
Sequence
Synthetic peptide located within the following region: SSFGSEAKAQQQEEQGSVNDVKEEEKEEKEVLPDQVEEEEENDDQEEEEE
Physical form
Lyophilized from PBS buffer with 2% sucrose
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Molecular and cellular biology, 30(18), 4404-4414 (2010-07-22)
Ribosome biogenesis is a highly regulated process ensuring that cell growth (increase in biomass) is coordinated with cell proliferation. The formation of eukaryotic ribosomes is a multistep process initiated by the transcription and processing of rRNA in the nucleolus. Concomitant
Molecular biology of the cell, 32(20), ar3-ar3 (2021-07-29)
Accumulating evidence indicates that increased ribosome biogenesis is a hallmark of cancer. It is well established that inhibition of any steps of ribosome biogenesis induces nucleolar stress characterized by p53 activation and subsequent cell cycle arrest and/or cell death. However
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.