Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV34545

Sigma-Aldrich

Anti-C14ORF166 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

C14ORF166 (CLE) is known to interact with influenza virus polymerase and HCV (Hepatitis C Virus) core protein during viral replication and assembly.
Rabbit Anti-C14ORF166 antibody recognizes bovine, human, mouse, rat, and canine C14ORF166.

Immunogen

Synthetic peptide directed towards the N terminal region of human C14orf166

Application

Rabbit Anti-C14ORF166 antibody is suitable for western blot applications at a concentration of 0.125 μg/ml.

Biochem/physiol Actions

The function of the C14orf266 gene has not yet been determined.

Sequence

Synthetic peptide located within the following region: GLAVRLEYGDNAEKYKDLVPDNSKTADNATKNAEPLINLDVNNPDFKAGV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Rodriguez et al.
Journal of virology, 85(22), 12062-12066 (2011-09-09)
The influenza A virus polymerase associates with a number of cellular transcription-related factors, including RNA polymerase II. We previously described the interaction of influenza virus polymerase subunit PA with human CLE/C14orf166 protein (hCLE), a positive modulator of this cellular RNA
Jun-Wei Lee et al.
Journal of proteome research, 10(10), 4522-4534 (2011-08-10)
The hepatitis C virus core protein (HCVc) forms the viral nucleocapsid and is involved in viral persistence and pathogenesis, possibly by interacting with host factors to modulate viral replication and cellular functions. Here, we identified 36 cellular protein candidates by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service