Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV34315

Sigma-Aldrich

Anti-FREQ antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Frequenin homolog (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

22 kDa

species reactivity

rat, human, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FREQ(23413)

General description

FREQ (NCS1) is a neuronal calcium sensor that modulates G protein-coupled receptor (GPCR) phosphorylation and synaptic functions. Interaction between variation in the DRD2 and FREQ genes can be used to predict the efficacy of nicotine replacement therapy.
Rabbit Anti-FREQ antibody recognizes bovine, human, mouse, rat, chicken, zebrafish FREQ.

Immunogen

Synthetic peptide directed towards the N terminal region of human FREQ

Application

Rabbit Anti-FREQ antibody is suitable for western blot (0.5 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

FREQ is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. This protein is thought to be associated with secretory granules and may be involved in the regulation of neurosecretion.

Sequence

Synthetic peptide located within the following region: MGKSNSKLKPEVVEELTRKTYFTEKEVQQWYKGFIKDCPSGQLDAAGFQK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Dahl et al.
The pharmacogenomics journal, 6(3), 194-199 (2006-01-13)
We have previously demonstrated that a functional dopamine D2 receptor promoter variant (DRD2 -141 Ins/Del) predicts response to nicotine replacement therapy (NRT). The present study extends this finding in the same population of 363 NRT-treated subjects, by examining variation in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service