Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

AV34163

Sigma-Aldrich

Anti-SIAH1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Seven in absentia homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$484.00

$484.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$484.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

guinea pig, bovine, mouse, rabbit, rat, dog, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIAH1(6477)

General description

SIAH1 is an E3 ligase that regulates protein ubiquitination and proteasome-mediated degradation. Studies have reported that the N-terminal RING domain regulates proteolysis, whereas the C-terminal sequences modulate target protein binding activities. It has also been implicated in p53 response-mediated degradation of β-catenin.
Rabbit Anti-SIAH1 (AB1) antibody recognizes bovine, human, mouse, rat, canine, and zebrafish SIAH1.

Immunogen

Synthetic peptide directed towards the N terminal region of human SIAH1

Application

Rabbit Anti-SIAH1 (AB1) antibody can be used for western blot applications at 1.25 μg/ml.

Biochem/physiol Actions

SIAH1 is a protein that is a member of the seven in absentia homolog (SIAH) family. The protein is an E3 ligase and is involved in ubiquitination and proteasome-mediated degradation of specific proteins. The activity of this ubiquitin ligase has been implicated in the development of certain forms of Parkinson′s disease, the regulation of the cellular response to hypoxia and induction of apoptosis. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.

Sequence

Synthetic peptide located within the following region: FTCLPAARTRKRKEMSRQTATALPTGTSKCPPSQRVPALTGTTASNNDLA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Hu et al.
Molecular and cellular biology, 19(1), 724-732 (1998-12-22)
The Drosophila seven in absentia (sina) gene was initially discovered because its inactivation leads to R7 photoreceptor defects. Recent data indicate that Sina binds to the Sevenless pathway protein Phyllopod, and together they mediate degradation of Tramtrack, a transcriptional repressor
S I Matsuzawa et al.
Molecular cell, 7(5), 915-926 (2001-06-08)
Destruction of beta-catenin is regulated through phosphorylation-dependent interactions with the F box protein beta-TrCP. A novel pathway for beta-catenin degradation was discovered involving mammalian homologs of Drosophila Sina (Siah), which bind ubiquitin-conjugating enzymes, and Ebi, an F box protein that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service