Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV33904

Sigma-Aldrich

Anti-KIF5B (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Kinesin family member 5B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product AV33904 is not currently sold in your country. Contact Technical Service

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

110 kDa

species reactivity

horse, mouse, rabbit, bovine, dog, rat, human, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIF5B(3799)

General description

Kinesin-1 heavy chain (KIF5B) is a member of the kinesin family of microtubule motors which aids in the movement of cargo along microtubules.

Immunogen

Synthetic peptide directed towards the C terminal region of human KIF5B

Biochem/physiol Actions

Kinesin is the founding member of a superfamily of microtubule based motor proteins that perform force-generating tasks such as organelle transport and chromosome segregation. Kinesin consists of heavy and light chains both of which have been documented to bind a variety of potential linker or cargo proteins. KIF5B is an isoform of kinesin heavy chain.

Sequence

Synthetic peptide located within the following region: ANEVKQAVEQQIQSHRETHQKQISSLRDEVEAKAKLITDLQDQNQKMMLE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

April M Morton et al.
Biochemical and biophysical research communications, 391(1), 388-393 (2009-11-17)
The cellular molecular motor kinesin-1 mediates the microtubule-dependent transport of a range of cargo. We have previously identified an interaction between the cargo-binding domain of kinesin-1 heavy chain KIF5B and the membrane-associated SNARE proteins SNAP-25 and SNAP-23. In this study
Ruibai Luo et al.
The Journal of biological chemistry, 291(41), 21350-21362 (2016-08-18)
The molecular basis for control of the cytoskeleton by the Arf GTPase-activating protein AGAP1 has not been characterized. AGAP1 is composed of G-protein-like (GLD), pleckstrin homology (PH), Arf GAP, and ankyrin repeat domains. Kif2A was identified in screens for proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service