Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV33747

Sigma-Aldrich

Anti-FOXQ1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Forkhead box Q1, Anti-HFH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

41 kDa

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

immunogen sequence

KLEVFVPRAAHGDKQGSDLEGAGGSDAPSPLSAAGDDSLGSDGDCAANSP

NCBI accession no.

UniProt accession no.

Storage temp.

−20°C

Gene Information

human ... FOXQ1(94234)

Immunogen

synthetic peptide corresponding to a region of human FOXQ1 with an internal ID of P23827

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jon Christensen et al.
PloS one, 8(3), e60051-e60051 (2013-04-05)
The forkhead box transcription factor FOXQ1 has been shown to be upregulated in colorectal cancer (CRC) and metastatic breast cancer and involved in tumor development, epithelial-mesenchymal transition and chemoresistance. Yet, its transcriptional regulation is still unknown. FOXQ1 mRNA and protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service