Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV32574

Sigma-Aldrich

Anti-MKX (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Mohawk homeobox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MKX(283078)

General description

MKX is a homeobox protein that belongs to the IRX family. This protein may modulate the development of tendons in mice.
Rabbit Anti-MKX (AB2) antibody recognizes zebrafish, human, chicken, canine, rat, bovine, and mouse MKX.

Immunogen

Synthetic peptide directed towards the middle region of human MKX

Application

Rabbit Anti-MKX can be used for western blot (0.25μg/ml) and IHC (4-8μg/ml) applications.

Biochem/physiol Actions

MKX contains 1 homeobox DNA-binding domain and belongs to the TALE/IRO homeobox family. It may act as a morphogenetic regulator of cell adhesion.

Sequence

Synthetic peptide located within the following region: IKSENSVIKAGVRPESRASEDYVAPPKYKSSLLNRYLNDSLRHVMATNTT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service