Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV32498

Sigma-Aldrich

Anti-CIC antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Capicua homolog (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

164 kDa

species reactivity

horse, mouse, bovine, rat, guinea pig, canine, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CIC(23152)

General description

CIC, a human homolog of Drosophila capicua, encodes a high mobility group box (HMG-box) transcription factor that is key sensor of receptor tyrosine kinase (RTK) signaling in Drosophila and mammals. CIC functions as a repressor of RTK-responsive genes, keeping them silent in the absence of signaling. CIC is susceptible to fusion to a double homeodomain gene DUX4 as a result of a recurrent chromosomal translocation, wherein the fusion product upregulates PEA3 family genes in Ewing-like sarcomas. Mutations in CIC are believed to contribute human oligodendroglioma.
Rabbit polyclonal anti-CIC reacts with human, mouse, rat, bovine, and canine capicua homolog (Drosophila) proteins.

Immunogen

The immunogen for anti-CIC antibody: synthetic peptide derected towards the N terminal of human CIC

Application

Rabbit Anti-CIC antibody can be used for IHC (4-8μg/ml, using paraffin-embedded tissues) and western blot (1.25μg/ml) applications.
Rabbit polyclonal anti-CIC is used to tag capicua homolog (Drosophila) for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of capicua homolog (Drosophila) in the regulation of RTK-promoted gene expression and various cancers such as Ewing-like sarcomas and oligodendrogliomas.

Sequence

Synthetic peptide located within the following region: PSQPDPSVQPSEAQQPASHPVASNQSKEPAESAAVAHERPPGGTGSADPE

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Gleize et al.
Annals of neurology, 78(3), 355-374 (2015-05-29)
CIC gene is frequently mutated in oligodendroglial tumors with 1p19q codeletion. However, clinical and biological impact remain poorly understood. We sequenced the CIC gene on 127 oligodendroglial tumors (109 with the 1p19q codeletion) and analyzed patients' outcome. We compared magnetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service