Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV32107

Sigma-Aldrich

Anti-FOXO1 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FKH1, Anti-FKHR, Anti-FOXO1A, Anti-Forkhead box O1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

dog, mouse, horse, rat, rabbit, human, bovine, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXO1(2308)

General description

Forkhead box O1/forkhead in rhabdomyosarcoma (FOXO1, FKH1, FKHR, FOXO1A), a forkhead transcription factor, is a preadipocye and myogenic differentiation factor and embryonic stem cell (ESC) maintenance factor that regulates gluconeogenesis and glycogenolysis by insulin signaling.
Rabbit polyclonal anti-FOXO1 antibody reacts with bovine, pig, canine, human, mouse, rat, zebrafish, and chicken Forkhead box O1/forkhead in rhabdomyosarcoma transcription factors.

Immunogen

Synthetic peptide directed towards the C terminal region of human FOXO1

Application

Rabbit Anti-FOXO1 (AB2) antibody can be used for western blot applications at 1μg/ml.
Rabbit polyclonal anti-FOXO1 antibody is used to tag Forkhead box O1/forkhead in rhabdomyosarcoma for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of Forkhead box O1/forkhead in rhabdomyosarcoma in cell differentiation, ESC maintenance and processes such as gluconeogenesis and glycogenolysis.

Biochem/physiol Actions

This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma.This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma.This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: HPMQMSALGGYSSVSSCNGYGRMGLLHQEKLPSDLDGMFIERLDCDMESI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Zhang et al.
Journal of diabetes research, 2019, 2583047-2583047 (2019-04-20)
Recent studies showed that alpha cells, especially immature cells and proalpha cells, might be the precursors of beta cells. Exposure to glucagon-like peptide 1 (GLP1) can ameliorate hyperglycemia in diabetic mice and restore the beta cell mass. In the present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service