Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV31981

Sigma-Aldrich

Anti-BARHL2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BarH-like 2 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

guinea pig, horse, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BARHL2(343472)

General description

BARHL2 is a homeobox transcription factor that regulates neuronal dI1 subtype diversification in the developing spinal cord.
Rabbit Anti-BARHL2 antibody recognizes rat, human, and mouse BARHL2.

Immunogen

Synthetic peptide directed towards the middle region of human BARHL2

Application

Rabbit Anti-BARHL2 antibody can be used for western blot applications at a concentration of 0.05-2.0μg/ml. It can also be used for IHC assays at 4-8μg/ml, using paraffin-embedded tissues.

Biochem/physiol Actions

BARHL2 and BARHL1 are two homeobox genes in mouse and human, which are highly related to the Bar Drosophila genes.

Sequence

Synthetic peptide located within the following region: SSPHHTPKQESNAVHESFRPKLEQEDSKTKLDKREDSQSDIKCHGTKEEG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Ding et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(5), 1566-1571 (2012-02-07)
In the dorsal spinal cord, distinct interneuron classes relay specific somatosensory information, such as touch, heat, and pain, from the periphery to higher brain centers via ipsilateral and contralateral axonal pathways. The transcriptional mechanisms by which dorsal interneurons choose between
Nobuhiko Tachibana et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(36), 9454-9471 (2016-09-09)
All tissues are genetically programmed to acquire an optimal size that is defined by total cell number and individual cellular dimensions. The retina contains stereotyped proportions of one glial and six neuronal cell types that are generated in overlapping waves.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service