Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

AV31834

Sigma-Aldrich

Anti-NR5A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Nuclear receptor subfamily 5, group A, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR5A1(2516)

General description

NR5A1 (or steroidogenic factor 1) is a nuclear receptor that modulates the transcription of genes involved in the hypothalamic-pituitary-steroidogenic axis. Mutations in NR5A1 have been linked to ovarian insufficiency and congenital adrenal hypoplasia.
Rabbit Anti-NR5A1 antibody recognizes bovine, canine, human, mouse, rat, and pig NR5A1.

Immunogen

Synthetic peptide directed towards the middle region of human NR5A1

Application

Rabbit Anti-NR5A1 antibody can be used for western blot (2.0μg/ml) and immunohistochemistry (4-8μg/ml, using paraffin embedded tissues) assays.

Biochem/physiol Actions

NR5A1 is an important regulator of steroidogeneisis which is present in human skin and its appendages. It plays a role in regulating p450scc expression with TReP-132 and CBP/p300. The protein encoded by this gene is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this gene are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: AVPGAHGPLAGYLYPAFPGRAIKSEYPEPYASPPQPGLPYGYPEPFSGGP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Lourenço et al.
The New England journal of medicine, 360(12), 1200-1210 (2009-02-28)
The genetic causes of nonsyndromic ovarian insufficiency are largely unknown. A nuclear receptor, NR5A1 (also called steroidogenic factor 1), is a key transcriptional regulator of genes involved in the hypothalamic-pituitary-steroidogenic axis. Mutation of NR5A1 causes 46,XY disorders of sex development
J K Phelan et al.
Human mutation, 18(6), 472-487 (2001-12-19)
Adrenal hypoplasia congenita (AHC) causes primary adrenal insufficiency due to the failure of development of the adrenal cortex. Clinical and pedigree data indicate that the condition is genetically heterogeneous. The predominant adrenal hypoplasia congenita locus, however, is the NR0B1 gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service