Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV31676

Sigma-Aldrich

Anti-ZNFN1A2 antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$457.00

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

43 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNFN1A2(22807)

General description

Helios/IKAROS family zinc finger 2 (ANF1A2, ZNF1A2, ZNFN1A2), a member of the Ikaros family of zinc-finger transcription factors, is a hematopoietic-specific transcription factors involved in the regulation of early lymphocyte development. Helios can form homodimers and heterodimers with other Ikaros family members to differentially regulate hematopoietic development.
Rabbit polyclonal anti-ZNFN1A2 antibody reacts with human, mouse, rat, chicken, and canine Helios/IKAROS family zinc finger 2 transcription factors.
Studies have reported that ZNFN1A2 has been deregulated in human leukemias.

Immunogen

Synthetic peptide directed towards the C terminal region of human ZNFN1A2

Application

Rabbit anti-ZNFN1A2 can be used for western blot (5.0μg/ml) and immunohistochemistry (4-8μg/ml, using paraffin-embedded tissues) applications.
Rabbit polyclonal anti-ZNFN1A2 antibody is used to tag helios/IKAROS family zinc finger 2 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of helios/IKAROS family zinc finger 2 in early onset regulation of lymphocyte development.

Biochem/physiol Actions

Novel short isoforms of this gene, ZNFN1A2, are overexpressed in a patient with T-cell acute lymphoblastic leukemia and may contribute to the development of T-cell malignancies.

Sequence

Synthetic peptide located within the following region: REASPSNSCLDSTDSESSHDDHQSYQGHPALNPKRKQSPAYMKEDVKALD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelita Rebollo et al.
Immunology and cell biology, 81(3), 171-175 (2003-05-20)
Ikaros, Aiolos and Helios encode zinc finger transcription factors that are important regulators of lymphoid development and differentiation. These proteins are involved in the control of gene expression and when associated with nuclear complexes, participate in nucleosome remodeling. Because differential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service