Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV31514

Sigma-Aldrich

Anti-SALF antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Stonin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

132 kDa

species reactivity

rabbit, human, bovine, dog, mouse, guinea pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SALF(11036)

General description

Rabbit polyclonal anti-SALF antibody reacts with human, mouse, zebrafish, canine, bovine, and rat stoning-1 clathrin adaptor factors.
Stonins are evolutionarily conserved, clathrin adaptor complex (AP-2mu-related) factors. They may function as cargo-specific sorting adaptors during cellular endocytosis. Stonin 1 (STON1 or SALF) is the human homolog of the Drosophila protein, stoned B.

Immunogen

Synthetic peptide directed towards the C terminal region of human SALF

Application

Rabbit Anti-SALF antibody can be used for western blot application at a concentration of 5μg/ml.
Rabbit polyclonal anti-SALF antibody is used to tag stonin-1 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of stonin-1 in processes related to cargo-specific sorting in endocytosis.

Biochem/physiol Actions

The SALF mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring SBLF and ALF genes. This rare transcript encodes a fusion protein composed of greater than 95% each of the individual elements, stoned B-like factor (SBLF) and TFIIA-alpha/beta-like factor (ALF). The significance of this co-transcribed mRNA and the function of its protein product have not yet been determined.

Sequence

Synthetic peptide located within the following region: NSGDDVSEQDVPDLFDTDNVIVCQYDKIHRSKNKWKFYLKDGVMCFGGRD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanja Maritzen et al.
Traffic (Copenhagen, Denmark), 11(1), 8-15 (2009-09-08)
Stonins are a small family of evolutionarily conserved clathrin adaptor complex AP-2mu-related factors that may act as cargo-specific sorting adaptors in endocytosis and perhaps beyond. Whereas little is known about the localization and function of stonin 1, recent work suggests

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service