AV30313
Anti-PDPK1 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
Anti-3-Phosphoinositide dependent protein kinase-1, Anti-MGC20087, Anti-MGC35290, Anti-PDK1, Anti-PRO0461, Anti-PkB-like, Anti-PkB-like1
Select a Size
$493.00
Select a Size
About This Item
$493.00
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
63 kDa
species reactivity
bovine, horse, dog, rabbit, human, mouse, guinea pig, rat
concentration
0.5 mg - 1 mg/mL
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PDPK1(5170)
General description
Rabbit Anti-PDPK1 antibody recognizes human, mouse, rat, zebrafish, and pig PDPK1.
Immunogen
Application
Biochem/physiol Actions
Sequence
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service