Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV13058

Sigma-Aldrich

Anti-RAB3D antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-D2-2, Anti-GOV, Anti-RAB16, Anti-RAB3D, member RAS oncogene family, Anti-RAD3D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB3D(9545)

General description

RAB3D, member RAS oncogene family (RAB3D, RAB16, RAD3D, GOV, D2-2) is a low molecular weight Ras-like GTPase involve in the regulation of intravesicle transport during exocytosis and apically directed transcytosis. RAB3D is found primarily in the secretory granules of non-neuronal exocrine secretory cells such as mast cells and pancreatic acinar cells.
Rabbit polyclonal anti-RAB3D antibody reacts with bovine, human, mouse, and rat RAB3D, member RAS oncogene family GTPases.

Immunogen

Synthetic peptide directed towards the C terminal region of human RAB3D

Application

Rabbit polyclonal anti-RAB3D antibody is used to tag RAB3D, member RAS oncogene family for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of RAB3D, member RAS oncogene family in exocytosis and transcytosis within non-neuronal secretory cells. Anti- RAB3D antibody produced in rabbit is suitable for western blotting at a concentration of 2.5 μg/ml.

Sequence

Synthetic peptide located within the following region: KENINVKQVFERLVDVICEKMNESLEPSSSSGSNGKGPAVGDAPAPQPSS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service