Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV100920

Sigma-Aldrich

Anti-E2F5 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

human, dog, rabbit, guinea pig, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... E2F5(1875)

General description

The members of E2F family of transcription factors bind to promoter regions and regulate cell proliferation.

Immunogen

Synthetic peptide directed towards the N terminal region of human E2F5

Application

Anti-E2F5 antibody produced in rabbit is suitable for western blotting at a concentration of 0.4 μg/ml.

Biochem/physiol Actions

Overexpression of E2F5 has been reported in solid tumors of breast, ovarian, colon and hepatocellular carcinoma. It is a transcription repressor with regulatory roles in cell proliferation, growth and cell cycle progression.

Sequence

Synthetic peptide located within the following region: KAEIEDLELKERELDQQKLLLQQSIKNVMDDSINNRFSYVTHEDICNCFN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzhu Jiang et al.
World journal of gastroenterology, 17(4), 470-477 (2011-01-29)
To explore the expression pattern of E2F5 in primary hepatocellular carcinomas (HCCs) and elucidate the roles of E2F5 in hepatocarcinogenesis. E2F5 expression was analyzed in 120 primary HCCs and 29 normal liver tissues by immunohistochemistry analysis. E2F5-small interfering RNA was
Alicia Subtil-Rodríguez et al.
Nucleic acids research, 42(4), 2185-2196 (2013-11-23)
The precise regulation of S-phase-specific genes is critical for cell proliferation. How the repressive chromatin configuration mediated by the retinoblastoma protein and repressor E2F factors changes at the G1/S transition to allow transcription activation is unclear. Here we show ChIP-on-chip

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service