Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV100755

Sigma-Aldrich

Anti-CREB5 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-cAMP responsive element binding protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

rat, human, pig, rabbit, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREB5(9586)

Immunogen

Synthetic peptide directed towards the N terminal region of human CREB5

Application

Anti-CREB5 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

CREB5 is a transcription factor, overexpressed in frontal cortex of HIV encephalitis patients. It is involved in critical biological processes of nucleic acid metabolism. It also regulates mRNA transcription and intracellular signaling cascades.

Sequence

Synthetic peptide located within the following region: CSLEHEFRKAQEEESSKRNISMHNAVGGAMTGPGTHQLSSARLPNHDTNV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Wang et al.
Cell biochemistry and biophysics, 60(3), 199-207 (2010-12-07)
CREB5 computational regulation network construction and analysis of frontal cortex of HIV encephalitis (HIVE) is very useful to identify novel markers and potential targets for prognosis and therapy. By integration of gene regulatory network infer and the database for annotation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service