Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV100591

Sigma-Aldrich

Anti-NANOG (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Nanog homeobox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NANOG(79923)

Immunogen

Synthetic peptide directed towards the N terminal region of human NANOG

Application

Anti-NANOG (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

Nanog is a transcription factor involved in the self-renewal maintainance of pluripotency of embryonic stem cells. It has important role in cell motility, epithelial-mesenchymal transition, proliferation and drug resistance.

Sequence

Synthetic peptide located within the following region: PMPVICGPEENYPSLQMSSAEMPHTETVSPLPSSMDLLIQDSPDSSTSPK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mong-Lien Wang et al.
OncoTargets and therapy, 6, 1207-1220 (2013-09-18)
The involvement of stemness factors in cancer initiation and progression has drawn much attention recently, especially after the finding that introducing four stemness factors in somatic cells is able to reprogram the cells back to an embryonic stem cell-like state.
Guangjin Pan et al.
Cell research, 17(1), 42-49 (2007-01-11)
Several extrinsic signals such as LIF, BMP and Wnt can support the self-renewal and pluripotency of embryonic stem (ES) cells through regulating the "pluripotent genes." A unique homeobox transcription factor, Nanog, is one of the key downstream effectors of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service