Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV07049

Sigma-Aldrich

Anti-CCL18 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Chemokine (C-C motif) ligand 18 (pulmonary and activation-Regulated)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

10 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

Storage temp.

−20°C

Gene Information

human ... CCL18(6362)

Immunogen

The immunogen for anti-CCL18 antibody: synthetic peptide derected towards the middle region of human CCL18

Application

Anti-CCL18 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

CCL18 is a weak chemoattractant for naïve T cells and recruits T and B lymphocytes and NK cells. It binds with a subset of CCR3 receptor population, CCR1, CCR2, CCR4 and CCR5, and inhibits chemotaxis.[1]

Sequence

Synthetic peptide located within the following region: PQKFIVDYSETSPQCPKPGVILLTKRGRQICADPNKKWVQKYISDLKLNA

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja C Krohn et al.
PloS one, 8(8), e72321-e72321 (2013-08-21)
CCL18 has been reported to be present constitutively at high levels in the circulation, and is further elevated during inflammatory diseases. Since it is a rather poor chemoattractant, we wondered if it may have a regulatory role. CCL18 has been
Sonja Krohn et al.
Frontiers in immunology, 4, 193-193 (2013-07-23)
The CC chemokine ligand 18 (CCL18) was first identified as a chemoattractant for naïve T cells. It has been reported to recruit T and B lymphocytes, and we show here, natural killer (NK) cells, but with low efficacy. Investigation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service