Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV07032

Sigma-Aldrich

Anti-CXCL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, α)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

8 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCL1(2919)

Immunogen

Synthetic peptide directed towards the middle region of human CXCL1

Application

Anti-CXCL1 antibody produced in rabbit is suitable for western blotting at a concentration of 0.0758 μg/ml.

Biochem/physiol Actions

CXCL1 is a chemokine that binds to CXCR2 receptor and stimulates migration of asthmatic airway smooth muscle cells. Prostate stromal cells secrete CXCL1 in response to IL-1 that results in prostatic inflammation in early stages of prostate cancer formation.

Sequence

Synthetic peptide located within the following region: QSVNVKSPGPHCAQTEVIATLKNGRKACLNPASPIVKKIIEKMLNSDKSN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ira Kogan-Sakin et al.
Carcinogenesis, 30(4), 698-705 (2009-02-24)
It is well accepted that tumor microenvironment is essential for tumor cells survival, cancer progression and metastasis. However, the mechanisms by which tumor cells interact with their surrounding at early stages of cancer development are largely unidentified. The aim of
Laila A Al-Alwan et al.
Journal of immunology (Baltimore, Md. : 1950), 191(5), 2731-2741 (2013-08-02)
Structural cell migration plays a central role in the pathophysiology of several diseases, including asthma. Previously, we established that IL-17-induced (CXCL1, CXCL2, and CXCL3) production promoted airway smooth muscle cell (ASMC) migration, and consequently we sought to investigate the molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service