ARLRCTB08A
PolarSafe™ Chest Freezer Rack
for 96 well microtube boxes, holds 8 boxes
All Photos(1)
About This Item
UNSPSC Code:
41122806
NACRES:
NB.22
material
stainless steel
manufacturer/tradename
Argos RCTB08A
General description
Argos PolarSafe Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2”, 3”, and 5” boxes, microtiter and deepwell plates and 15 & 50 mL centrifuge tubes.
Argos PolarSafe Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright or Chest ultra low freezers.
Argos PolarSafe Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright or Chest ultra low freezers.
Legal Information
PolarSafe is a trademark of Argos Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)