Key Documents
ARLP0534
Plastic Pasteur Pipette
L 5 3/4 in., sterile, polystyrene
About This Item
UNSPSC Code:
41121510
NACRES:
NB.22
material
polystyrene
sterility
sterile
manufacturer/tradename
Argos
packaging
pkg of individually wrapped, 50/bag, 200/case
L
5 3/4 in.
General description
Argos plastic Pasteur pipets are produced from medical grade polystyrene in a class 100,000 cleanroom. Plastic Pasteur pipets offer the same aspirating precision of glass, and the safety and benefits of plastic. Plastic construction allows the long flexible stem to be bent to draw liquid from narrow or small volume tubes. These pipets can be used with aspirating bulbs or vacuum applications to remove liquid waste from flasks, dishes, plates & tubes.
Pipets are available in two sizes (5.75" or 9") and are supplied individually wrapped to maintain sterility.
Choose from one of the most recent versions:
Certificates of Analysis (COA)