Key Documents
ARLB8801BL
PolarSafe Ice pan
capacity 1 L, black polystyrene
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
material
black polystyrene
manufacturer/tradename
Argos B8801BL
capacity
1 L
General description
Argos PolarSafe Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2”, 3”, and 5” boxes, microtiter and deepwell plates and 15 & 50 mL centrifuge tubes.
Argos PolarSafe Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright or Chest ultra low freezers.
Argos Technologies is pleased to introduce a family of high density expanded polystyrene ice buckets and pans.
Our containers are produced with thick walls to provide rigidity and strength, and yet are super lightweight. These new PolarSafe storage products are perfectly suited for use with ice, dry ice, slurry ice, etc.
The 3 pans and 2 buckets provide excellent insulating properties to help keep temperature sensitive samples safely chilled, but will not leave condensation on the benchtop. We purposely designed our containers for stable and secure benchtop use, but also added features to make them easy to carry from lab to lab. Our buckets come with snug fitting lids to minimize evaporation loss and to maximize cooling.
The PolarSafe buckets and pans are cool containers that are a less costly alternative to expensive and cumbersome polyurethane models.
Legal Information
PolarSafe is a trademark of Argos Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)