MilliporeSigma
All Photos(4)
Key Documents
ARLAPS2
Serological Pipettes
capacity 2 mL, sterile, polystyrene
About This Item
UNSPSC Code:
41106514
NACRES:
NB.22
material
polystyrene
sterility
sterile
feature
individually wrapped
packaging
pkg of 500 ea
manufacturer/tradename
Argos APS2
capacity
2 mL
General description
Individually wrapped plastic pipettes that are supplied without graduations or filters for vacuum or free flow pipetting applications. Can also be used with peristaltic pumps.
Choose from one of the most recent versions:
Certificates of Analysis (COA)