Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AQ0928

AQUA Peptide INSVEVYDGTLY[R]

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116026
Pricing and availability is not currently available.

mol wt

monoisotopic mol wt 1538.71 Da

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ptk7(71461)

Application

Protein-AQUA is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope–labeled peptides and mass spectrometry.

Legal Information

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.
AQUA is a trademark of Harvard Medical School

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service