form
solid
mol wt
monoisotopic mol wt 1059.517 Da
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... XIRP1(165904) , XIRP2(129446)
mouse ... Xirp1(22437) , Xirp2(241431)
rat ... Cmya1_predicted(316071) , Cmya3(311098)
Application
Protein-AQUA™ is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope–labeled peptides and mass spectrometry.
Legal Information
This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.
This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
AQUA is a trademark of Harvard Medical School
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service