Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AQ0097

Sigma-Aldrich

AQUA Peptide EAVI[F]K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2513C9H55N615NO9
Molecular Weight:
715.77
UNSPSC Code:
12352200
NACRES:
NA.26

form

solid

mol wt

monoisotopic mol wt 715.433 Da

shipped in

wet ice

storage temp.

−20°C

Application

Protein AQUA is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope–labeled peptides and mass spectrometry.

Legal Information

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.
This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company.  The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration.  The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party.  Information about licenses for excluded uses is available from:  E. I. du Pont de Nemours and Company;  Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
AQUA is a trademark of Harvard Medical School
Protein AQUA is a trademark of Harvard Medical School

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service