APREST88305
PrEST Antigen CFAP46
Prestige Antigens™ Powered by Atlas Antibodies, buffered aqueous solution
Synonym(s):
C10orf123, C10orf124, C10orf92, C10orf93, DKFZp434A1721, FLJ25954, TTC40, bA288G11.4, bA288G11.5, bB137A17.2, bB137A17.3
About This Item
recombinant
expressed in E. coli
assay
>80% (SDS-PAGE)
form
buffered aqueous solution
mol wt
predicted mol wt 28 kDa
purified by
immobilized metal affinity chromatography (IMAC)
concentration
≥0.5 mg/mL
immunogen sequence
LKRRMVLAQQYLAQASEVLLQCLQVALGSGLLDVAAAASLEMVECVGTLDPATTCQFLALSQSCSASETMRDVLLAATANTSSSQLAALLQLQHQLR
Ensembl | human accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CFAP46(54777)
General description
Application
Linkage
Physical form
Preparation Note
Legal Information
Disclaimer
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service