Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APREST76982

Sigma-Aldrich

PrEST Antigen AKNAD1

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):

C1orf62, MGC26989

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in E. coli

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 26 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

EEEAIIKSIISCYNKNSWPKEQTPELTDQLNPKRDGENSNKPGSATTTEENTSDLEGPVAAGDSSHQENVNVLTKTKGP

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AKNAD1(254268)

General description

Recombinant protein fragment of Human AKNAD1 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.

Application

Suitable as a blocking agent using corresponding antibodies.

Linkage

Corresponding Antibody HPA030270.

Physical form

Solution in 1 M urea-PBS, pH 7.4

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service