Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APREST74099

Sigma-Aldrich

PrEST Antigen GABRQ

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):

THETA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in E. coli

tag

His tagged

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 30 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

DGVSSLPITPAQAPLASPESLGSLTSTSEQAQLATSESLSPLTSLSGQAPLATGESLSDLPSTSEQARHSYGVRFNGFQADDSIFPTEIRNRVEAHGHGVTHDHEDSNESLSSDE

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GABRQ(55879)

General description

Recombinant protein fragment of Human GABRQwith N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.

Application

Blocking agent and positive assay control using corresponding antibodies.

Linkage

Corresponding Antibodies HPA002063

Physical form

Solution in phosphate-buffered saline and 1M Urea, pH 7.4

Preparation Note

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service