Key Documents
ADT60112-EP
Adapter for Eppendorf® T-60-11 Rotor
holds 10 x 1.5 – 2 mL reaction vessels, pack of 6 ea
Select a Size
6 EA
$685.00
$685.00
Check Cart for Availability
Select a Size
Change View
6 EA
$685.00
About This Item
UNSPSC Code:
41103911
$685.00
Check Cart for Availability
feature
holds 10 x 1.5 – 2 mL reaction vessels
packaging
pack of 6 ea
manufacturer/tradename
Eppendorf® 022638521 (NA)
parameter
16,400 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with Rotor T-60-11
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)