Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0365

Sigma-Aldrich

Claudin-7 (EP399) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$897.00

$897.00


Estimated to ship onJuly 17, 2025FromSAINT LOUIS


Request a Bulk Order

Select a Size

Change View
1 ML
$897.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$897.00


Estimated to ship onJuly 17, 2025FromSAINT LOUIS


Request a Bulk Order

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

General description

The claudin family of epithelial tight junction proteins that play critical roles for cell-cell adhesion and maintenance of cellular polarity. Claudin members are expressed in a tissue specific manner; claudin-7 expression is localized to the distal nephron epithelium. Claudin-7 has been extensively studied in renal cell carcinoma (RCC) subtypes. By immunohistochemical (IHC) analysis, membranous claudin-7 expression was detected in 67% of chromophobe RCC, compared with 0% of clear cell RCCs, 28% of papillary RCCs, and 23% oncocytomas. Claudin-7 expression may provide clinical utility in the differential diagnosis of chromophobe RCC and oncocytoma. While both chromophobe RCC and oncocytoma are derived from intercalated cells of the cortical collecting duct, oncocytomas are typically benign lesions whereas chromophobe RCCs are potentially aggressive malignancies. These RCC subtypes can be difficult to differentiate by morphology alone. Hornsby and colleagues examined claudin-7 IHC and reported 71% and 73% positive and negative predictive values for the differential diagnosis of chromophobe RCC from oncocytoma.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &lt0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service