Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0350A

Sigma-Aldrich

Neuron-specific enolase (NSE) (EP319) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
$301.00

$301.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 ML
$301.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$301.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Neuron-specific enolase (NSE), also known as enolase 2 (ENO2), is a 78 kDa gamma-enolase homodimer and one of three mammalian isozymes catalyzing the interconversion of 2-phosphoglycerate and phosphoenolpyruvate. The C-terminus promotes cell survival by regulating neuronal growth factor receptor-dependent signaling pathways. NSE localizes in the cytosol and is expressed in mature neurons and throughout the neuroendocrine system, predominantly in cells of the amine precursor uptake and decarboxylation lineage. Literature reports showed low expression of NSE in other cell types, where it may be located on the membrane and have non-glycolytic functions. These include erythrocyte, platelet, prostate, uterine, breast, striated muscle, and smooth muscle cells. However, this antibody specifically labels neuronendocrine cells in normal tissues. NSE is the most sensitive immunohistochemical marker for middle to late stage small cell lung cancer (SCLC) and can also identify Merkel cells and melanocytes. NSE overexpression is primarily found in tumors of neurogenic and neuroendocrine origin. Incidences of normally NSE-negative cells producing NSE have been found in malignant transformations; thus, NSE should be paired with the more specific bombesin, which does not stain non-neuroendocrine cells, and/or chromogranin, which intensely stains lung carcinoid tumors. Serum NSE is also useful in identifying small-cell lung carcinoma (SCLC), carcinoids, islet cell tumors and neuroblastomas, and are potential indicators of neural injury, such as reactive gliosis, astrocytic death, and blood-brain barrier dysfunction.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and & < 0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service