Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0349

Sigma-Aldrich

CD248 (Endosialin) (EP383) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$897.00

$897.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 ML
$897.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$897.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

storage temp.

2-8°C

visualization

(membrane,cytoplasm)

General description

CD248, known also as endosialin and tumor endothelial marker 1 (TEM1), is a 95-kDa transmembrane glycoprotein expressed in activated stromal and perivascular fibroblasts and pericytes. CD248 is an important regulator of critical pathways involved with stromal fibroblast migration, and proliferation. During normal embryonic development, CD248 is highly expressed across diverse tissue types. Postnatally, its expression is restricted to the endometrium, bone marrow and corpus luteum. In vitro protein structure studies demonstrated interaction of the extracellular domain of CD248 with extracellular matrix proteins, facilitating cell migration and metastatic formation. Conversely, the cytoplasmic domain was shown to regulate stromal fibroblast function. CD248 is expressed within the endothelium across many tumor types include sarcomas, carcinoma and neuroectodermal tumors. Within the tumor stroma, activated fibroblasts subpopulations are often associated with poor prognosis.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1%BSA and &< 0.1%Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service