Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0344A

Sigma-Aldrich

GRIA2 (EP387) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
$301.00

$301.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 ML
$301.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$301.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

visualization

(cytoplasm/membrane)

General description

Glutamate receptor 2 (GRIA2) is a ligand-gated ion channel that uses L-glutamate for excitatory synaptic transmission expressed in the central nervous system. A recent gene expression profiling study reported >100 fold GRIA2 induction in solitary fibrous tumors compared with control tissues. Immunohistochemistry studies demonstrated GRIA2 positivity in the majority of solitary fibrous tumors (64-86%). Its diagnostic performance yields 64% sensitivity, 92% specificity, with 41% and 97% positive and negative predictive value, respectively for the differential diagnosis of solitary fibrous tumors versus other mesenchymal soft tissue tumors. GRIA2 may be a useful marker to identify STAT6-negative solitary fibrous tumors.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &< 0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service