Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0062

Sigma-Aldrich

CD61 (GPIIIa) (EP65) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,050.00

$1,050.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$1,050.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$1,050.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

Ig fraction of antiserum

clone

monoclonal

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

storage temp.

2-8°C

visualization

(membrane/cytoplasm)

General description

CD61, also known as GPIIIa and Integrin beta-3, is a glycoprotein encoded by the ITGB3 gene. CD61 heterodimerize with other cluster of differentiation molecules resulting in transmembrane integrins, facilitating cell to cell interactions and signal transduction. CD61 combines with CD41 to form platelet glycoprotein IIb/IIIa that serves as a receptor for fibrinogen and fibronectin. Platelet activation causes a conformational change in the glycoprotein to bind soluble fibrinogen to from a platelet plug. Alternative, CD61 can heterodimerize with CD51 to form Integrin alpha V beta 3, a receptor for vitronectin and von Willebrand factor. CD61 is expressed in the membrane and cytoplasm of platelets and megakaryocytes across diverse tissue types. Endothelium is also stained. Most cases of acute megakaryoblastic leukemia (AML M7) are positive for CD61.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &lt0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service