Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A9893

Sigma-Aldrich

N6-(6-Aminohexyl)adenosine 2′,5′-diphosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N6O10P2 · xLi+
CAS Number:
Molecular Weight:
526.38 (free acid basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

assay

>90% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Li+].NCCCCCCNc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3OP(O)(O)=O

InChI

1S/C16H28N6O10P2.Li/c17-5-3-1-2-4-6-18-14-11-15(20-8-19-14)22(9-21-11)16-13(32-34(27,28)29)12(23)10(31-16)7-30-33(24,25)26;/h8-10,12-13,16,23H,1-7,17H2,(H,18,19,20)(H2,24,25,26)(H2,27,28,29);/q;+1/p-1/t10-,12-,13-,16-;/m1./s1

InChI key

BZVXLSAPAPOGFA-KHXPSBENSA-M

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service