Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A9354

Sigma-Aldrich

Ac-Arg-Glu(EDANS)-Val-His-His-Gln-Lys-Leu-Val-Phe-Lys(DABCYL)-Arg-OH trifluoroacetate salt

≥95% (HPLC), powder

Synonym(s):

Ac-RE(EDANS)-VHHQKLVF-K(DABCYL)-R-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C100H144N30O20S · xC2HF3O2
Molecular Weight:
2118.47 (free base basis)
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥95% (HPLC)

form

powder

storage condition

desiccated
protect from light
under inert gas

color

red to purple

shipped in

wet ice

storage temp.

−20°C

Amino Acid Sequence

Ac-Arg-Glu-EDANS-Val-His-His-Gln-Lys-Leu-Val-Phe-Lys-DABCYL-Arg

Biochem/physiol Actions

α-Secretase substrate II, fluorogenic.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Komano et al.
The Journal of biological chemistry, 273(48), 31648-31651 (1998-11-21)
Human beta-amyloid precursor protein (APP) introduced into yeast undergoes alpha-secretase-type cleavage, suggesting that yeast have alpha-secretase-like protease(s). Here we report that two structurally and functionally related glycosyl-phosphatidylinositol-linked yeast aspartyl proteases, Mkc7p and Yap3p (collectively termed yapsin), are responsible for alpha-secretase-type

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service