Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A9295

Sigma-Aldrich

4-Androsten-4-ol-3,17-dione acetate

Synonym(s):

4-Acetoxy-4-androstene-3,17-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
form:
solid

form

solid

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(=O)OC1=C2CC[C@H]3[C@@H]4CCC(=O)[C@@]4(C)CC[C@@H]3[C@@]2(C)CCC1=O

InChI

1S/C21H28O4/c1-12(22)25-19-16-5-4-13-14-6-7-18(24)21(14,3)10-8-15(13)20(16,2)11-9-17(19)23/h13-15H,4-11H2,1-3H3/t13-,14-,15-,20+,21-/m0/s1

InChI key

LRXSFNGKRCOHRS-VMRCMBGLSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of aromatase inhibitor on oestrogen production in rabbit blastocysts.
J T Wu et al.
Journal of reproduction and fertility, 66(2), 655-662 (1982-11-01)
Aromatase enzyme catalysis is involved in the potent inhibition of estrogen biosynthesis caused by 4-acetoxy- and 4-hydroxy-4-androstene-3, 17-dione.
D F Covey et al.
Molecular pharmacology, 21(1), 173-180 (1982-01-01)
A M Brodie
Cancer research, 42(8 Suppl), 3312s-3314s (1982-08-01)
Since the first publication in 1973 concerning aromatase inhibitors, several effective compounds have been reported by a number of investigators. Our studies with 4-hydroxyandrostene-3,17-dione, 4-acetoxyandrostene-3,17-dione, and 1,-4,6-androstatrienedione indicate that these compounds cause rapid competitive inhibition of the enzyme. Aminoglutethimide binds
M Motta et al.
Journal of steroid biochemistry, 25(4), 593-600 (1986-10-01)
The present study reports the effects exerted by 1,4,6-androstatriene-3,17-dione (ATD), 4-hydroxy-4-androstene-3,17-dione (4-OH-A) and 4-acetoxy-4-androstene-3,17-dione (4-Ac-A), three steroids known to inhibit the aromatization of androgens to estrogens, on the in vitro metabolism of labelled testosterone (T), dihydrotestosterone (DHT) and androstenedione (delta-4-A)
J T Wu
Journal of reproduction and fertility, 72(2), 345-349 (1984-11-01)
The ability of aromatase inhibitors to induce implantation in mice was tested in animals in which implantation was delayed by ovariectomy and progesterone treatment. Implantation was consistently induced by 7 mg 4-hydroxyandrostene-3,17-dione (4-OH-A), 7 X 5 mg 1,4,6-androstatriene-3,17-dione (ATD) or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service