Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8606

Sigma-Aldrich

Anti-α2C Adrenergic Receptor antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-α-2-adrenergic receptor, renal type, Anti-ADRA2C, Anti-ADRA2L2, Anti-ALPHA2CAR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:100-1:300
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRA2C(152)
mouse ... Adra2c(11553)
rat ... Adra2c(24175)

General description

Adrenergic receptors are a G-protein coupled receptors that exist in two forms, α or β. Ligands for adrenergic receptors are catecholamines that induce a sympathetic response in animal cells. [1]
Studies have reported that α2C adrenergic receptors can regulate adrenergic opioid synergy and spinal analgesia. Rabbit Anti-α2C Adrenergic Receptor antibody binds to humanα2C Adrenergic Receptor.

Immunogen

synthetic peptide corresponding to an internal sequence of human α2C Adrenergic Receptor.

Application

Rabbit Anti-α2C Adrenergic Receptor antibody can be used for immunohistochemistry (1:100-1:300) and western blot (1:500-1:1,000) applications.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolyn A Fairbanks et al.
The Journal of pharmacology and experimental therapeutics, 300(1), 282-290 (2001-12-26)
The alpha(2A)-adrenergic receptor (AR) subtype mediates antinociception induced by the alpha(2)AR agonists clonidine, dexmedetomidine, norepinephrine, and 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine (UK-14,304) as well as antinociceptive synergy of UK-14,304 with opioid agonists [D-Ala(2),N-Me-Phe(4),Gly(5)-ol]-enkephalin and deltorphin II. Differential localization of alpha(2)-adrenergic (alpha(2A)-, alpha(2B)-(,) alpha(2C)-) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service