A8474
StripSpin™ adapter
can be easily attached and removed from the rotor with one hand, pkg of 1 ea
Synonym(s):
equipment replacement accessory
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Labnet
technique(s)
cryopreservation: suitable
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service