A801000
Polystyrene Support 80 dA(bz)
Synonym(s):
dA(bz) Polystyrene column
About This Item
Recommended Products
type
for DNA synthesis
product line
Proligo Reagents
assay
≥97.0% (Nucleoside purity, reversed phase HPLC)
λ
conforms (UV/VIS Identity)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service