Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7851

Sigma-Aldrich

Monoclonal Anti-Hamster IgG−Peroxidase antibody produced in mouse

clone MAH 1.12, purified immunoglobulin, lyophilized powder

Synonym(s):

Monoclonal Anti-Hamster IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

MAH 1.12, monoclonal

form

lyophilized powder

species reactivity

hamster

should not react with

rat, bovine, horse, chicken, goat, human, mouse, pig, rabbit

technique(s)

direct ELISA: 1:40,000 using hamster IgG as the coating protein
western blot: 1:2,000 using recombinant mouse TNFα

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Hamster IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-hamster IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Mouse Monoclonal Anti-hamster IgG-Peroxidase antibody is immunospecific for hamster IgG and does not recognize human, bovine, horse, goat, pig, rabbit, rat, mouse, and chicken IgG.

Immunogen

purified Armenian hamster monoclonals (TN3-19.2 and H22).

Application

Mouse Monoclonal Anti-Hamster IgG-Peroxidase antibody is suitable for ELISA (1:40,000) and western blotting (1:2,000) applications.
Parasite cell lysates were analyzed by western blot using goat anti-hamster IgG at a 1:5000 dilution for 1 hour at room temperature.

Physical form

Lyophilized powder containing 0.01 M phosphate buffered saline, pH 7.4, 1% bovine serum albumin, and 0.01% merthiolate.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly Campbell et al.
Infection and immunity, 72(4), 2194-2202 (2004-03-25)
Several Leishmania proteins have been identified and characterized in pursuit of understanding pathogenesis and protection in cutaneous leishmaniasis. In the present study, we utilized sera from infected BALB/c mice to screen a Leishmania amazonensis amastigote cDNA expression library and obtained
Hong Guo et al.
Oncology letters, 14(1), 383-389 (2017-07-12)
The soluble urokinase-type plasminogen activator receptor (suPAR) and the urokinase-type plasminogen activator receptor (uPAR) have been proposed as useful biomarkers of tumor progression. Recently, suPAR was associated with chemoresistance in lung cancer. However, its clinical significance in leukemia has not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service