Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

A7734

Sigma-Aldrich

Anti-ADFP (31-45) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ADRP, Anti-Adipophilin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADFP(123)

General description

ADFP is a member of the perilipin family of proteins. It is associated with lipid surface membrane material.
Rabbit Anti-ADFP (31-45) antibody binds to human ADFP (31-45).

Immunogen

synthetic peptide corresponding to amino acids 31-45 of human ADFP

Application

Rabbit Anti-ADFP (31-45) antibody can be used for western blot applications at dilutions ranging from 1:500 to 1:2,000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pia Kotokorpi et al.
Molecular pharmacology, 77(1), 79-86 (2009-10-22)
Expression of adipocyte differentiation-related protein (ADFP), residing on the surface of lipid droplets, correlates to hepatic fat storage. In the context of consequences and treatment of metabolic disorders, including hepatic steatosis, it is imperative to gain knowledge about the regulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service