Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7707

Eppendorf® PCR adapter plates

Synonym(s):

96 well pcr plates, pcr adapter plates, pcr plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.76

sterility

non-sterile

feature

lid: no
adapter plate designed to insert into exchangeable thermoblock for microtiter plates (MTPs)

packaging

case of 1
pack of 1

manufacturer/tradename

Eppendorf® 022670573 (USA)
Eppendorf® 5363 007.009 (EU)

technique(s)

PCR: suitable

size

96 wells

color

none

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® PCR Adapter Plates

Special rack designed for use with the thermoblock microplate (T3942); accommodates 0.2 mL PCR tubes, 1 x 8 strip tubes and 96 well microplates.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service