Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7602

Sigma-Aldrich

Anti-ADAM-33, Cytoplasmic Domain antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A Disintegrin And Metalloproteinase-33

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM33(80332)

General description

A Disintegrin And Metalloproteinase-33 (ADAM-3) is a widely expressed protein that regulates cell-attachment by binding to integrins via cysteine-rich domains. ADAM-33 has also been associated with asthma susceptibility. Rabbit Anti-ADAM-33, Cytoplasmic Domain antibody binds to the cytoplasmic end of human ADAM-33.
By immunoblotting against the reduced protein, the antibody identifies bands at 99 kDa (minor band), 58 kDa (major band), and cleaved products in cell lysates.

Immunogen

synthetic peptide corresponding to cytoplasmic domain of human ADAM-33.

Application

Rabbit Anti-ADAM-33, Cytoplasmic Domain antibody can be used for western blot assays at 1:1,000.

Physical form

Solution in phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Van Eerdewegh et al.
Nature, 418(6896), 426-430 (2002-07-12)
Asthma is a common respiratory disorder characterized by recurrent episodes of coughing, wheezing and breathlessness. Although environmental factors such as allergen exposure are risk factors in the development of asthma, both twin and family studies point to a strong genetic
Tsuyoshi Yoshinaka et al.
Gene, 282(1-2), 227-236 (2002-01-30)
The ADAM family of membrane-anchored proteins has a unique domain structure, with each containing a disintegrin and metalloprotease (ADAM) domain. We have isolated mouse and human cDNAs encoding a novel member of the ADAM family. The mouse and human predicted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service