Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7590

Sigma-Aldrich

Adenosine-2,8-3H 5′-monophosphate diammonium salt

≥95% (Radiochemical Purity, HPLC), aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N7O7P
Molecular Weight:
381.28
MDL number:
UNSPSC Code:
41106305

Quality Level

assay

≥95% (Radiochemical Purity, HPLC)

form

aqueous ethanol solution

extent of labeling

10-30 Ci/mmol

storage temp.

−20°C

SMILES string

[P](=O)([O-])([O-])OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n]2c3nc(nc(c3nc2[3H])N)[3H].[N+H4].[N+H4]

InChI key

DGXWJGDBZBXMIT-CLFYFHHFSA-N

Packaging

Combi-vial

Physical form

Ethanol: water (1:1) solution

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service