Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7465

Sigma-Aldrich

Adenosine-8-14C cyclic monophosphate ammonium salt

≥95% (HPLC), aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106305
NACRES:
NA.28

Quality Level

assay

≥95% (HPLC)

form

aqueous ethanol solution

extent of labeling

40-60 mCi per mmol

shipped in

dry ice

storage temp.

−70°C

SMILES string

[P]1(=O)(O[C@@H]2[C@H](O[C@H]([C@@H]2O)[n]3[14cH]nc4c3ncnc4N)CO1)[O-].[N+H4]

InChI key

LURMYAAZBGGMPK-GNDWCDBVSA-N

Packaging

Combi-vial

Physical form

Ethanol: water (1:1) solution

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

80.6 °F - closed cup

flash_point_c

27 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service