ADAM 13 belongs to the metalloproteinase family having a disintegrin-like domain (ADAM). ADAM13 may modulate cell-attachment by associating with integrins through cysteine-rich domains. It is also involved in the migration of neural crest-cells. Rabbit Anti-ADAM-13, Propeptide Region antibody binds to Xenopus ADAM13.
By immunoblotting against the reduced protein, the antibody identifies bands at 120 kDa (minor band), 92 kDa (major band), and cleaved products at 68 kDa, 40 kDa in cell lysates.
Immunogen
synthetic peptide corresponding to the amino end of Xenopus ADAM-13.
Application
Rabbit Anti-ADAM-13, Propeptide Region antibody can be used for western blot applications at a dilution of 1:1,000.
Physical form
Solution in phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.
Current biology : CB, 11(12), 918-930 (2001-07-13)
Cranial neural-crest (CNC) cells originate from the lateral edge of the anterior neuroepithelium and migrate to form parts of the peripheral nervous system, muscles, cartilage, and bones of the face. Neural crest-cell migration involves the loss of adhesion from the
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.